Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANGRF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118236
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118236 Supplier Invitrogen™ Supplier No. PA5118236
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RANGRF Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

May regulate the intracellular trafficking of RAN. In cardiac cells seems to regulate the cell surface localization of SCN5A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANGRF
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene RANGRF
Gene Accession No. Q9HD47
Gene Alias 2400006H24Rik; HSPC165; HSPC236; MDS5; MOG1; MOG1 homolog; ran guanine nucleotide release factor; Ran-binding protein MOG1; Rangnrf; RANGRF
Gene Symbols RANGRF
Host Species Rabbit
Immunogen E. coli-derived Human RANGRF fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29098
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.