Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANK (CD265) Monoclonal Antibody (D13), Invitrogen™

Catalog No. PIMA543900
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543900 20 μL
PIMA543899 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543900 Supplier Invitrogen™ Supplier No. MA543900
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RANK is a member of the tumor necrosis factor receptor (TNFR) family. The full length mouse RANK cDNA encodes a type I transmembrane protein with an extracellular and a cytoplasmic domain. The extracellular domain contains two potential N-linked glycosylation sites. RANK shares significant amino acid homology with other members of the TNFR family in its extracellular four cysteine-rich repeats. Human and murine RANK share 81% amino acid identity in their extracellular domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANK (CD265)
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone D13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TNFRSF11A
Gene Accession No. Q9Y6Q6
Gene Alias CD265; FEO; LOH18CR1; loss of heterozygosity, 18, chromosomal region 1; Ly109; mRANK; ODFR; OFE; OPTB7; Osteoclast differentiation factor receptor; OSTS; Paget disease of bone 2; PDB2; Rank; receptor activator of NF-kappaB; receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; RGD1563614; TNF receptor superfamily member 11a; Tnfrsf11a; TRANCER; TRANCE-R; tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Gene Symbols TNFRSF11A
Host Species Mouse
Immunogen Val330-Lys615 (Accession Q9Y6Q6), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498206, 8792
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.