Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RANK/TNFRSF11A Antibody (044), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290884H
Description
RANK/TNFRSF11A Monoclonal specifically detects RANK/TNFRSF11A in Rat samples. It is validated for ELISA.Specifications
RANK/TNFRSF11A | |
Monoclonal | |
HRP | |
CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat RANK/TNFRSF11A (XP_573424.2; Met1-Pro213). | |
0.1 mL | |
Cell Cycle and Replication | |
8792 | |
Rat | |
Purified |
ELISA | |
44 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Rat EDAR | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction