Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RANK/TNFRSF11A Antibody (107), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290324AF532
Description
RANK/TNFRSF11A Monoclonal specifically detects RANK/TNFRSF11A in Human samples. It is validated for ELISA.Specifications
RANK/TNFRSF11A | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
107 | |
ELISA | |
CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human RANK/TNFRSF11A (NP_001257879.1; Met1-Pro212). | |
0.1 mL | |
Cell Cycle and Replication | |
8792 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction