Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RANK/TNFRSF11A Antibody (9A725), HRP, R&D Systems™
SDP

Catalog No. NB10056508H Shop All Novus Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056508H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056508H Supplier R&D Systems Supplier No. NB10056508H
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

RANK/TNFRSF11A Monoclonal specifically detects RANK/TNFRSF11A in Avian, Human, Mouse, Rat samples. It is validated for CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen RANK/TNFRSF11A
Applications CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 9A725
Conjugate HRP
Dilution Western Blot, Immunohistochemistry-Paraffin
Gene Accession No. Q9Y6Q6
Gene Alias CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Gene Symbols TNFRSF11A
Host Species Mouse
Immunogen A fusion protein containing amino acid residues 326-616 of human RANK was used as immunogen. This antibody recognizes an epitope located between amino acid residues 330-427 of human RANK (NP_003830).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8792
Test Specificity In RAW cells, a 97 kDa is observed.
Target Species Avian, Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.