Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RANK/TNFRSF11A Antibody (9A725), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056508MFV450
Description
RANK/TNFRSF11A Monoclonal antibody specifically detects RANK/TNFRSF11A in Human, Mouse, Rat, Avian samples. It is validated for Flow Cytometry, Flow (Cell Surface)Specifications
RANK/TNFRSF11A | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Avian | |
Purified |
Flow Cytometry | |
9A725 | |
Flow Cytometry, Flow (Cell Surface) | |
CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
A fusion protein containing amino acid residues 326-616 of human RANK was used as immunogen. This antibody recognizes an epitope located between amino acid residues 330-427 of human RANK (NP_003830). | |
0.1 mL | |
Cell Cycle and Replication | |
8792 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction