Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RANK/TNFRSF11A Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen RANK/TNFRSF11A
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056396T
SDP
View Documents
Novus Biologicals
NB10056396SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056396
SDP
View Documents
Novus Biologicals
NB10056396
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

RANK/TNFRSF11A Polyclonal specifically detects RANK/TNFRSF11A in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

RANK/TNFRSF11A
Western Blot 1-3 ug/ml
Polyclonal
Purified
RUO
Human, Mouse
Q9Y6Q6
8792
A synthetic peptide corresponding to amino acid residues 603-613 of human RANK (NP_003830).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Cell Cycle and Replication
PBS containing 0.05% BSA with 0.05% Sodium Azide
CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator
TNFRSF11A
IgG
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.