Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RANK/TNFRSF11A Antibody (SR2241), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | RANK/TNFRSF11A |
---|---|
Clone | SR2241 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
RANK/TNFRSF11A Monoclonal antibody specifically detects RANK/TNFRSF11A in Human samples. It is validated for Western BlotSpecifications
RANK/TNFRSF11A | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
A synthesized peptide derived from human RANK/TNFRSF11A (Uniprot #: Q9Y6Q6) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2241 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
8792 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title