Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RANK/TNFRSF11A Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $526.00
Specifications
| Antigen | RANK/TNFRSF11A |
|---|---|
| Clone | 80710 |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB6832SP
![]() |
R&D Systems
MAB6832SP |
25 μg |
Each for $200.50
|
|
|||||
MAB6832100
![]() |
R&D Systems
MAB6832100 |
100 μg |
Each for $526.00
|
|
|||||
Description
RANK/TNFRSF11A Monoclonal antibody specifically detects RANK/TNFRSF11A in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
| RANK/TNFRSF11A | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| RANK/TNFRSF11A | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 80710 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9Y6Q6 | |
| Mouse myeloma cell line NS0-derived recombinant human RANK/TNFRSF11A, extracellular domain, Accession # Q9Y6Q6 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title