Learn More
Stoelting™ Rankin Forceps
SDP
Hemostatic forceps are used in surgical procedures to control bleeding or the flow of liquids in tubing.
$197.20
Specifications
Length (English) 6.25 in.
View More Specs
Products 2
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-001-336
SDP
View Documents
Stoelting™
5211018
Straight
Each for $197.20
Only null left
Add to Cart
10-001-337
SDP
View Documents
Stoelting™
5211019
Curved
Each for $197.20
Only null left
Add to Cart
Description
Description
  • Scissor-like ring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
Specifications
Specifications
6.25 in.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.