Learn More
RANKL (CD254) Monoclonal Antibody (IK22/5), Functional Grade, In Vivo PLATINUM™, Leinco Technologies™

Rat Monoclonal Antibody
Supplier: Leinco Technologies Inc R1368100MG


Description
Specificity: Clone IK22/5 recognizes an epitope on mouse RANKL. Endotoxin Level: ≤ 0.5 EU/mg as determined by the LAL method. Purity: ≥98% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco Purified Functional PLATINUM™ antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
RANKL (CD254) Monoclonal antibody specifically detects RANKL (CD254) in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays, Western BlotSpecifications
RANKL (CD254) | |
Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
O35235 | |
Rat | |
Protein A/G | |
RUO | |
21943 | |
4°C short term, -80°C long term | |
Liquid |
Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays, Western Blot | |
IK22/5 | |
Functional Grade | |
TNFSF11 | |
CD254; H-RANK-L; hRANKL2; Ly109l; M-Rank-L; ODF; OPG; OPG ligand; OPGL; OPTB2; Osteoclast differentiation factor; osteoprotegerin ligand; Rankl; rank-l; receptor activator of NF-kappaB ligand; receptor activator of nuclear factor kappa B ligand; receptor activator of nuclear factor kappa-B ligand; RP11-86N24.2; sOdf; srankl; TNF-related activation-induced cytokine; Tnfsf11; TNLG6B; Trance; tumor necrosis factor (ligand) superfamily member 11; tumor necrosis factor (ligand) superfamily, member 11; tumor necrosis factor ligand 6B; tumor necrosis factor ligand superfamily member 11; Tumor necrosis factor ligand superfamily member 11, membrane form; Tumor necrosis factor ligand superfamily member 11, soluble form; tumor necrosis factor superfamily member 11; tumor necrosis factor-related activation-induced cytokine | |
Purified Recombinant Mouse RANKL (NSO-derived) | |
100 mg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.