Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RANKL Mouse anti-Human, Unconjugated, Clone: Anti-UBE3A, clone 10H7.1, Cat. No. MABS1683, is a mouse monoclonal antibody that detects Ubiquitin-p, MilliporeSigma™

Catalog No. MABS169625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1696-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1696-25 Supplier MilliporeSigma Supplier No. MABS169625UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-RANKL, clone 6A12.1, Cat. No. MABS1696, is a mouse monoclonal antibody that detets RANKL in human cells and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANKL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Anti-UBE3A, clone 10H7.1, Cat. No. MABS1683, is a mouse monoclonal antibody that detects Ubiquitin-p
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Mouse monoclonal antibody in tissue culture supernatant without azide.
Gene Alias Tumor necrosis factor ligand superfamily member 11;Osteoclast differentiation factor;ODF;Osteoprotegerin ligand;OPGL;Receptor activator of nuclear factor kappa-B ligand;TNF-related activation-induced cytokine;CD254
Gene Symbols TNFSF11;OPGL;RANKL;TRANCE
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 95 amino acids from the C-terminal region of human RANKL.
Molecular Weight of Antigen ∽36 kDa observed; 35.48 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 6A12.1 detects RANKL protein in human cells. It targets an epitope within 95 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Product Type Antibodies
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.