Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RANKL Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA5110268

Catalog No. PIPA5110268


Description

Description

Sequence of this protein is as follows: RISEDGTHCI YRILRLHENA DFQDTTLESQ DTKLIPDSCR RIKQAFQGAV QKELQHIVGS QHIRAEKAMV DGSWLDLAKR SKLEAQPFAH LTINATDIPS GSHKVSLSSW YHDRGWAKIS NMTFSNGKLI VNQDGFYYLY ANICFRHHET SGDLATEYLQ LMVYVTKTSI KIPSSHTLMK GGSTKYWSGN SEFHFYSINV GGFFKLRSGE EISIEVSNPS LLDPDQDATY FGAFKVRDID

This gene encodes a member of the tumor necrosis factor (TNF) cytokine family which is a ligand for osteoprotegerin and functions as a key factor for osteoclast differentiation and activation. This protein was shown to be a dentritic cell survival factor and is involved in the regulation of T cell-dependent immune response. T cell activation was reported to induce expression of this gene and lead to an increase of osteoclastogenesis and bone loss. This protein was shown to activate antiapoptotic kinase AKT/PKB through a signaling complex involving SRC kinase and tumor necrosis factor receptor-associated factor (TRAF) 6, which indicated this protein may have a role in the regulation of cell apoptosis. Targeted disruption of the related gene in mice led to severe osteopetrosis and a lack of osteoclasts. The deficient mice exhibited defects in early differentiation of T and B lymphocytes, and failed to form lobulo-alveolar mammary structures during pregnancy. Two alternatively spliced transcript variants have been found.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RANKL
Polyclonal
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14788, O35235, Q9ESE2
TNFSF11
Recombinant fusion protein containing a sequence corresponding to amino acids 78-317 of human RANKL (NP_003692.1).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TNFSF11
CD254; H-RANK-L; hRANKL2; Ly109l; M-Rank-L; ODF; OPG; Opgl; OPTB2; Osteoclast differentiation factor; Osteoprotegerin ligand; Rankl; Receptor activator of nuclear factor kappa-B ligand; RP11-86N24.2; sOdf; TNF-related activation-induced cytokine; Tnfsf11; TNLG6B; Trance; Tumor necrosis factor ligand superfamily member 11; Tumor necrosis factor ligand superfamily member 11, membrane form; Tumor necrosis factor ligand superfamily member 11, soluble form
Rabbit
Antigen affinity chromatography
RUO
117516, 21943, 8600
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.