Learn More
RANKL (soluble) Polyclonal Antibody, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
$259.00 - $2255.00
Specifications
| Antigen | RANKL (soluble) |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P133G50
|
PeproTech
500P133G50UG |
50 μg |
Each for $259.00
|
|
|||||
500P133G100
|
PeproTech
500P133G100UG |
100 μg |
Each for $331.00
|
|
|||||
500P133G1MG
|
PeproTech
500P133G1MG |
1 mg |
Each for $2,255.00
|
|
|||||
Description
AA Sequence of recombinant protein: MEKAMVDGSW LDLAKRSKLE AQPFAHLTIN ATDIPSGSHK VSLSSWYHDR GWAKISNMTF SNGKLIVNQD GFYYLYANIC FRHHETSGDL ATEYLQLMVY VTKTSIKIPS SHTLMKGGST KYWSGNSEFH FYSINVGGFF KLRSGEEISI EVSNPSLLDP DQDATYFGAF KVRDID. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human sRANK Ligand. Anti-Human sRANK Ligand-specific antibody was purified by affinity chromatography employing an immobilized Human sRANK Ligand matrix. Sandwich ELISA: To detect Human sRANK Ligand by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human sRANK Ligand (500-P133GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human sRANK Ligand. Western Blot: To detect Human sRANK Ligand by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human sRANK Ligand is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
This gene encodes a member of the tumor necrosis factor (TNF) cytokine family which is a ligand for osteoprotegerin and functions as a key factor for osteoclast differentiation and activation. This protein was shown to be a dentritic cell survival factor and is involved in the regulation of T cell-dependent immune response. T cell activation was reported to induce expression of this gene and lead to an increase of osteoclastogenesis and bone loss. This protein was shown to activate antiapoptotic kinase AKT/PKB through a signaling complex involving SRC kinase and tumor necrosis factor receptor-associated factor (TRAF) 6, which indicated this protein may have a role in the regulation of cell apoptosis. Targeted disruption of the related gene in mice led to severe osteopetrosis and a lack of osteoclasts. The deficient mice exhibited defects in early differentiation of T and B lymphocytes, and failed to form lobulo-alveolar mammary structures during pregnancy. Two alternatively spliced transcript variants have been found.Specifications
| RANKL (soluble) | |
| ELISA, Western Blot | |
| Unconjugated | |
| Goat | |
| Human | |
| O14788 | |
| 8600 | |
| E.coli-derived Recombinant Human sRANK Ligand. | |
| Antigen affinity chromatography | |
| TNFSF11 |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| CD254; H-RANK-L; hRANKL2; Ly109l; M-Rank-L; ODF; OPG; OPG ligand; OPGL; OPTB2; Osteoclast differentiation factor; osteoprotegerin ligand; Rankl; rank-l; receptor activator of NF-kappaB ligand; receptor activator of nuclear factor kappa B ligand; receptor activator of nuclear factor kappa-B ligand; RP11-86N24.2; sOdf; srankl; TNF-related activation-induced cytokine; Tnfsf11; TNLG6B; Trance; tumor necrosis factor (ligand) superfamily member 11; tumor necrosis factor (ligand) superfamily, member 11; tumor necrosis factor ligand 6B; tumor necrosis factor ligand superfamily member 11; Tumor necrosis factor ligand superfamily member 11, membrane form; Tumor necrosis factor ligand superfamily member 11, soluble form; tumor necrosis factor superfamily member 11; tumor necrosis factor-related activation-induced cytokine | |
| TNFSF11 | |
| Primary | |
| -20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.