Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145050
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145050 Supplier Invitrogen™ Supplier No. PA5145050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ANLTDKELEA FREELKHFEA KIEKHNHYQK QLEIAHEKLR HAESVGDGER VSRSREKHAL LEGRTKELGY TVKKHLQDLS GRISRARHNE L Antigen sequence identity: Mouse 78%; Rat 78%.

The term RAP typically refers to receptor-associated protein, encoded by the LRPAP1 gene, which plays a significant role in the trafficking and function of certain receptors within cells. RAP is primarily involved in the regulation of low-density lipoprotein receptor-related proteins (LRPs) by acting as a chaperone during protein transport within the endoplasmic reticulum (ER). It prevents premature ligand binding and assists in the proper folding and assembly of receptor complexes, ensuring efficient receptor function and trafficking to the cell surface. RAP also influences cellular processes related to cholesterol metabolism and cell signaling through its interactions with these receptors. Its function is crucial for transporting and recycling receptors involved in various pathways, including lipid metabolism, endocytosis, and cellular homeostasis. Dysregulation of RAP or related receptor functions can impact metabolic health and are implicated in conditions such as atherosclerosis, neurodegenerative disorders, and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lrpap1
Gene Accession No. P30533
Gene Alias 39 kDa receptor-associated protein; A2MRAP; A2RAP; AA617339; AI790446; Alpha-2-macroglobulin receptor-associated protein; Alpha-2-macroglobulin receptor-associated protein precursor (Alpha-2-MRAP) (Low density lipoprotein receptor-related protein-associated protein 1) (RAP) (Heparin binding protein-44) (HBP-44); alpha-2-MRAP; AU042172; C77774; gp330-binding 45 kDa protein; HBP44; HBP-44; heparin-binding protein 44; LDL receptor related protein associated protein 1; low density lipoprotein receptor related protein, associated protein 1; low density lipoprotein receptor-related protein associated protein 1; low density lipoprotein receptor-related protein-associated protein 1; low density lipoprotein-related protein-associated protein 1 (alpha-2-macroglobulin receptor-associated protein 1); Lrpap1; MRAP; MYP23; RAP
Gene Symbols Lrpap1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 267-357. Recombinant protein control fragment (Product #RP-110154).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4043
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.