Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580778
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human LRPAP1.
The term RAP typically refers to receptor-associated protein, encoded by the LRPAP1 gene, which plays a significant role in the trafficking and function of certain receptors within cells. RAP is primarily involved in the regulation of low-density lipoprotein receptor-related proteins (LRPs) by acting as a chaperone during protein transport within the endoplasmic reticulum (ER). It prevents premature ligand binding and assists in the proper folding and assembly of receptor complexes, ensuring efficient receptor function and trafficking to the cell surface. RAP also influences cellular processes related to cholesterol metabolism and cell signaling through its interactions with these receptors. Its function is crucial for transporting and recycling receptors involved in various pathways, including lipid metabolism, endocytosis, and cellular homeostasis. Dysregulation of RAP or related receptor functions can impact metabolic health and are implicated in conditions such as atherosclerosis, neurodegenerative disorders, and cancer.
Specifications
RAP | |
Polyclonal | |
Unconjugated | |
Lrpap1 | |
39 kDa receptor-associated protein; A2MRAP; A2RAP; AA617339; AI790446; Alpha-2-macroglobulin receptor-associated protein; Alpha-2-macroglobulin receptor-associated protein precursor (Alpha-2-MRAP) (Low density lipoprotein receptor-related protein-associated protein 1) (RAP) (Heparin binding protein-44) (HBP-44); alpha-2-MRAP; AU042172; C77774; gp330-binding 45 kDa protein; HBP44; HBP-44; heparin-binding protein 44; LDL receptor related protein associated protein 1; low density lipoprotein receptor related protein, associated protein 1; low density lipoprotein receptor-related protein associated protein 1; low density lipoprotein receptor-related protein-associated protein 1; low density lipoprotein-related protein-associated protein 1 (alpha-2-macroglobulin receptor-associated protein 1); Lrpap1; MRAP; MYP23; RAP | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
4043 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with no preservative | |
P30533 | |
Lrpap1 | |
A synthetic peptide corresponding to the center region of the Human LRPAP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction