Learn More
Invitrogen™ RAP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588927
Description
Immunogen sequence: YSREKNQPKP SPKRESGEEF RMEKLNQLWE KAQRLHLPPV RLAELHADLK IQERDELAWK KLKLDGLDED GEKEARLIRN LNVILAKYGL DGKKDARQVT SNSLSGTQED GLDDPRLEKL WHKAKTSGKF SGEELDKLWR EFLHHKEKVH EYNVLLETLS RTEEIHENVI SPSDLSDIKG SVLHSRHTEL KEKLRSINQG LDRLRRVSHQ GYSTEAEFEE PRVIDLWDLA QSANLTDKEL EAFREELKHF EAKIEKHNHY QKQLEIAHEK LRHAESVGDG ERVSRSREKH ALLEGRTKEL GYTVKKHLQD LSGRISRARH NEL; Positive Samples: 293T, HeLa, A-375; Cellular Location: Cell surface, Cytoplasm, Endoplasmic reticulum.
The term RAP typically refers to receptor-associated protein, encoded by the LRPAP1 gene, which plays a significant role in the trafficking and function of certain receptors within cells. RAP is primarily involved in the regulation of low-density lipoprotein receptor-related proteins (LRPs) by acting as a chaperone during protein transport within the endoplasmic reticulum (ER). It prevents premature ligand binding and assists in the proper folding and assembly of receptor complexes, ensuring efficient receptor function and trafficking to the cell surface. RAP also influences cellular processes related to cholesterol metabolism and cell signaling through its interactions with these receptors. Its function is crucial for transporting and recycling receptors involved in various pathways, including lipid metabolism, endocytosis, and cellular homeostasis. Dysregulation of RAP or related receptor functions can impact metabolic health and are implicated in conditions such as atherosclerosis, neurodegenerative disorders, and cancer.
Specifications
RAP | |
Polyclonal | |
Unconjugated | |
Lrpap1 | |
39 kDa receptor-associated protein; A2MRAP; A2RAP; AA617339; AI790446; Alpha-2-macroglobulin receptor-associated protein; Alpha-2-macroglobulin receptor-associated protein precursor (Alpha-2-MRAP) (Low density lipoprotein receptor-related protein-associated protein 1) (RAP) (Heparin binding protein-44) (HBP-44); alpha-2-MRAP; AU042172; C77774; gp330-binding 45 kDa protein; HBP44; HBP-44; heparin-binding protein 44; LDL receptor related protein associated protein 1; low density lipoprotein receptor related protein, associated protein 1; low density lipoprotein receptor-related protein associated protein 1; low density lipoprotein receptor-related protein-associated protein 1; low density lipoprotein-related protein-associated protein 1 (alpha-2-macroglobulin receptor-associated protein 1); Lrpap1; MRAP; MYP23; RAP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
116565, 16976, 4043 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.83 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P30533, P55302, Q99068 | |
Lrpap1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 35-357 of human LRPAP1 (NP_0023281). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.