Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575367 Supplier Thermo Scientific Supplier No. PA575367
Add to Cart
Add to Cart

RAP1 Polyclonal Antibody

RAP1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene TERF2IP
Gene Accession No. Q5EAN7, Q91VL8, Q9NYB0
Gene Alias dopamine receptor interacting protein 5; dopamine receptor-interacting protein 5; DRIP5; hRap1; MNCb-0448; MNCb-0628; PP8000; RAP1; RAP1 homolog; repressor/activator protein 1 homolog; tel; telomeric repeat binding factor 2, interacting protein; telomeric repeat-binding factor 2-interacting protein 1; TERF2 interacting protein; TERF21P; TERF2-interacting telomeric protein 1; TERF2IP; TRF2-interacting telomeric protein 1; TRF2-interacting telomeric protein Rap1; TRF2-interacting telomeric RAP1 protein
Gene Symbols TERF2IP
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 220-270 of Human RAP1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307861, 54386, 57321
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.