Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAP1A Antibody (1D2-1C64), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222526
Description
RAP1A Monoclonal specifically detects RAP1A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
RAP1A | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 2 ug, Immunohistochemistry-Paraffin 1:100 - 1:1000 | |
C21KG, G-22K, GTP-binding protein smg p21A, KREV-1, KREV1Ras-related protein Krev-1, RAP1, RAP1A, member of RAS oncogene family, ras-related protein Rap-1A, RAS-related protein RAP1A, SMGP21 | |
Mouse | |
21 kDa | |
100 μg | |
Cell Cycle and Replication | |
5906 | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1D2-1C64 | |
Unconjugated | |
P62834 | |
RAP1A | |
Recombinant full-length human Rap1 | |
Protein A purified | |
RUO | |
Primary | |
This product was made specifically to the RAP1A human protein (P62834). It has not yet been tested for cross reactivity to other related proteins. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction