Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rap1A/B Antibody (SR1333), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322136100UL
Description
Rap1A/B Monoclonal antibody specifically detects Rap1A/B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunoprecipitationSpecifications
Rap1A/B | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
5906 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunoprecipitation | |
SR1333 | |
Western Blot 1:1000-1:5000, Flow Cytometry 1:40, Immunoprecipitation 1:50 | |
RAP1A, RAP1B | |
A synthesized peptide derived from human Rap1A/B (Uniprot #: P61224, P62834) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction