Learn More
Invitrogen™ RAP1A Monoclonal Antibody (1D2-1C64)

Description
Western blot analysis of MA1-013 detects an ~21kDa protein.
Specifications
Specifications
| Antigen | RAP1A |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1D2-1C64 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol, 1mg/mL BSA and 0.05% sodium azide |
| Gene | RAP1A |
| Gene Accession No. | P62834, P62835 |
| Gene Alias | AI848598; C21KG; G-22K; GTP-binding protein; GTP-binding protein smg p21A; GTP-binding protein SMG-P21A; KREV1; Krev-1; rap 1A; RAP1; RAP1A; RAP-1A; RAP1A, member of RAS oncogene family; RAS-related protein 1a; ras-related protein Krev-1; Ras-related protein Rap-1A; RAS-related protein-1a; SMGP21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.