Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAP1A Monoclonal Antibody (1D4-1A3)
GREENER_CHOICE

Catalog No. PIMA1147
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1147 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1147 Supplier Invitrogen™ Supplier No. MA1147
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Western blot analysis of MA1-147 detects an ~21kDa protein. In IHC (paraffin) applications, this antibody has only shown reactivity with human samples.

The product of this gene belongs to the family of RAS-related proteins. These proteins share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. The most striking difference between RAP proteins and RAS proteins resides in their 61st amino acid: glutamine in RAS is replaced by threonine in RAP proteins. The product of this gene counteracts the mitogenic function of RAS because it can interact with RAS GAPs and RAF in a competitive manner. Two transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP1A
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1D4-1A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene RAP1A
Gene Accession No. P62834, P62835
Gene Alias AI848598; C21KG; G-22K; GTP-binding protein; GTP-binding protein smg p21A; GTP-binding protein SMG-P21A; KREV1; Krev-1; rap 1A; RAP1; RAP1A; RAP-1A; RAP1A, member of RAS oncogene family; RAS-related protein 1a; ras-related protein Krev-1; Ras-related protein Rap-1A; RAS-related protein-1a; SMGP21
Gene Symbols RAP1A
Host Species Mouse
Immunogen Recombinant full-length human Rap1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109905, 5906
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.