Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (6C7G0)
GREENER_CHOICE

Catalog No. PIMA538205
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538205 Supplier Invitrogen™ Supplier No. MA538205
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, NIH/3T3, C6, U-251MG, HUVEC, Rat lung, Rat spleen Immunogen sequence: LYMKNGQGFA LVYSITAQST FNDLQDLREQ ILRVKDTDDV PMILVGNKCD LEDERVVGKE QGQNLARQWN NCAFLESSAK S.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP1A/RAP1B
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 6C7G0
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAP1A
Gene Accession No. P61224, P62834, P62835, P62836, Q62636, Q99JI6
Gene Alias 2810443E11Rik; AI848598; C21KG; G-22K; GTP-binding protein smg p21A; GTP-binding protein smg p21B; GTP-binding protein SMG-P21A; K-REV; KREV1; Krev-1; OK/SW-cl.11; RAL1B; rap 1A; RAP1; RAP1A; RAP-1A; RAP1A, member of RAS oncogene family; Rap1b; RAP1B, member of RAS oncogene family; Ras family small GTP binding protein RAP1B; RAS related protein 1b; RAS-related protein 1a; Ras-related protein Krev-1; Ras-related protein Rap-1A; RAS-related protein RAP1B; ras-related protein Rap-1b; RAS-related protein-1a; small GTP binding protein; SMGP21
Gene Symbols RAP1A, RAP1B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 70-150 of human RAP1A + RAP1B (P61224).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109905, 171337, 215449, 295347, 5906, 5908
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.