Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (JE35-61)
GREENER_CHOICE

Catalog No. FEMA550823
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550823 Supplier Invitrogen™ Supplier No. MA550823
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: HeLa cell lysate, Jurkat cell lysate, NIH/3T3 cell lysate, HEK-293 cell lysate, HEK-293. Predicted band size: 20 kDa Subcellular Location: Cell membrane, Cytoplasm, Cytoplasm, perinuclear region, Cell junction, Early endosome; Cell membrane, Cytoplasm, cytosol, Cell junction.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP1A/RAP1B
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone JE35-61
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene RAP1B
Gene Accession No. P61224, P62834, P62835, Q99JI6
Gene Alias 2810443E11Rik; AI848598; C21KG; G-22K; GTP-binding protein smg p21A; GTP-binding protein smg p21B; GTP-binding protein SMG-P21A; K-REV; KREV1; Krev-1; OK/SW-cl.11; RAL1B; rap 1A; RAP1; RAP1A; RAP-1A; RAP1A, member of RAS oncogene family; Rap1b; RAP1B, member of RAS oncogene family; Ras family small GTP binding protein RAP1B; RAS related protein 1b; RAS-related protein 1a; Ras-related protein Krev-1; Ras-related protein Rap-1A; RAS-related protein RAP1B; ras-related protein Rap-1b; RAS-related protein-1a; small GTP binding protein; SMGP21
Gene Symbols RAP1A, RAP1B
Host Species Rabbit
Immunogen Synthetic peptide within Human RAP1A aa 93-142/184.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109905, 215449, 5906, 5908
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.