Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAP1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598268
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598268 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598268 Supplier Invitrogen™ Supplier No. PA598268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAP1B Polyclonal Antibody for Western Blot, IHC (P), ELISA

Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP1B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RAP1B
Gene Accession No. P61224, Q99JI6
Gene Alias 2810443E11Rik; cb1026; cb119; GTP-binding protein (smg p21B); GTP-binding protein smg p21B; K-REV; OK/SW-cl.11; RAL1B; rap1b; RAP1B, member of RAS oncogene family; Ras family small GTP binding protein RAP1B; RAS related protein 1b; ras-related protein rab-1B-like protein; RAS-related protein RAP1B; ras-related protein Rap-1b; Ras-related protein Rap-1b-like protein; sb:cb119; si:dkey-31f5.4; small GTP binding protein; wu:fb11c04; wu:fb74e09; wu:fk70e05
Gene Symbols RAP1B
Host Species Rabbit
Immunogen Recombinant Human Ras-related protein Rap-1b protein (1-181AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215449, 5908
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.