Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726645 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6645 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6645 Supplier Proteintech Group Inc Supplier No. 108401AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAP1B and RAP1A belong to a superfamily of RAS like small GTP-binding proteins involved in cell signaling.

Specifications

Antigen RAP1B
Applications Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RAP1B
Gene Accession No. P61224, Q62636, Q99JI6
Gene Alias GTP binding protein smg p21B, K REV, RAL1B, RAP1B, Ras related protein Rap 1b
Gene Symbols Rap1b
Host Species Rabbit
Immunogen RAP1B Fusion Protein Ag1282
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171337, 215449, 5908
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.