Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP1GAP Mouse anti-Human, Clone:2G7B6, Abnova™

Catalog No. 89333202 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-202 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-202 Supplier Abnova Corporation Supplier No. MAB16625
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human RAP1GAP.

Specifications

Antigen RAP1GAP
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2G7B6
Conjugate Unconjugated
Description RAP1 GTPase activating protein
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene RAP1GAP
Gene Alias KIAA0474, RAP1GA1, Rap1GAP1, rap1GAPII
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 412-678 of human RAP1GAP from E. coli.
Quantity 100 μg
Gene ID (Entrez) 5909
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.