Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAP1GAP Recombinant Rabbit Monoclonal Antibody (8V9I6)
GREENER_CHOICE

Catalog No. PIMA542831
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542831 Supplier Invitrogen™ Supplier No. MA542831
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: BxPC-3, U-87MG, Mouse brain, Rat brain. The target is usually found in the following locations: Golgi apparatus membrane, Peripheral membrane protein. Immunogen sequence: MIEKMQGSRM DEQRCSFPPP LKTEEDYIPY PSVHEVLGRE GPFPLILLPQ FGGYWIEGTN HEITSIPETE PLQSPTTKVK LECNPTARIY RKHFLGKEHF.

Rap1GAP specifically stimulates GTP hydrolytic activity of the monomeric G protein Rap1. Physical interaction between G alpha z, a member of the Gi family of trimeric G proteins, and Rap1GAP blocks the ability of regulators of G protein signaling to stimulate GTP hydrolysis of the alpha subunit, and also attenuates the ability of activated G alpha z to inhibit adenylyl cyclase. Rap1GAP is expressed in the brain, kidney and pancreas and may act as a signal integrator to coordinate and/or integrate Gz signaling and Rap1 signaling in cells. A novel isoform of Rapl GTPase-activating protein, designated Rap1GAPII, binds specifically to G alpha z. Stimulation of the Gi-coupled M2 Muscarinic receptor translocates Rap1GAPII from the cytosol to the membrane and decreases the amount of GTP-bound Rap1, resulting in the activation of ERK/MAPK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP1GAP
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 8V9I6
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAP1GAP
Gene Accession No. A2ALS5, P47736
Gene Alias 1300019I11Rik; 2310004O14Rik; AI427470; ARPP-90; KIAA0474; RAP1 GTPase activating protein; Rap1 GTPase-activating protein; rap1 GTPase-activating protein 1; Rap1, GTPase-activating protein 1; Rap1ga1; Rap1GAP; Rap1GAP1; RAP1GAPII; RAPGAP
Gene Symbols RAP1GAP
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Rap1GAP (P47736).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110351, 313644, 5909
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.