Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAP1GDS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528471
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528471 Supplier Invitrogen™ Supplier No. PA528471
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: PC-3. Predicted reactivity: Mouse (91%), Chicken (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The smg GDP dissociation stimulator (smgGDS) protein is a stimulatory GDP/GTP exchange protein with GTPase activity (Riess et al., 1993 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP1GDS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene RAP1GDS1
Gene Accession No. P52306
Gene Alias BC011279; Exchange factor smgGDS; GDS1; LOW QUALITY PROTEIN: rap1 GTPase-GDP dissociation stimulator 1; rap1 GTPase-GDP dissociation stimulator 1; rap1 GTPase-GDP dissociation stimulator 1; LOW QUALITY PROTEIN: rap1 GTPase-GDP dissociation stimulator 1; RAP1, GTP-GDP dissociation stimulator 1; RAP1GDS1; smg GDP/GTP exchange protein; SMG GDS protein; SMG P21 stimulatory GDP/GTP exchange protein; SmgGDS
Gene Symbols RAP1GDS1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human RAP1GDS1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5910
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.