Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP1GDS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545009
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545009 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545009 Supplier Thermo Scientific Supplier No. PA545009
Add to Cart
Add to Cart

RAP1GDS1 Polyclonal Antibody

RAP1GDS1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP1GDS1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RAP1GDS1
Gene Accession No. P52306
Gene Alias BC011279; Exchange factor smgGDS; GDS1; LOW QUALITY PROTEIN: rap1 GTPase-GDP dissociation stimulator 1; rap1 GTPase-GDP dissociation stimulator 1; rap1 GTPase-GDP dissociation stimulator 1; LOW QUALITY PROTEIN: rap1 GTPase-GDP dissociation stimulator 1; RAP1, GTP-GDP dissociation stimulator 1; RAP1GDS1; smg GDP/GTP exchange protein; SMG GDS protein; SMG P21 stimulatory GDP/GTP exchange protein; SmgGDS
Gene Symbols RAP1GDS1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RAP1GDS1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5910
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.