Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAP2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107231
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107231 Supplier Invitrogen™ Supplier No. PA5107231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%; Sheep: 93%; Zebrafish: 79%.

RAP2A is a small GTP-binding protein which cycles between a GDP-bound inactive and a GTP-bound active form. In its active form interacts with and regulates several effectors including MAP4K4, MINK1 and TNIK. Part of a signaling complex composed of NEDD4, RAP2A and TNIK which regulates neuronal dendrite extension and arborization during development. More generally, it is part of several signaling cascades and may regulate cytoskeletal rearrangements, cell migration, cell adhesion and cell spreading.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP2A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene RAP2A
Gene Accession No. P10114
Gene Alias 5830461H18Rik; KREV; K-REV; RAP2; RAP2, member of RAS oncogene family (K-rev); Rap2a; Rap2A GTPase; RAP2A, member of RAS oncogene family; RAS related protein 2a; ras-related protein Rap-2a; RbBP-30; RP11-128N14.2
Gene Symbols RAP2A
Host Species Rabbit
Immunogen Synthetic peptide located within the following region: QIIRVKRYEK VPVILVGNKV DLESEREVSS SEGRALAEEW GCPFMETSAK.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5911
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.