Learn More
Invitrogen™ RAP2A Polyclonal Antibody

Description
RAP2A Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | RAP2A |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | RAP2A |
| Gene Accession No. | P10114, Q80ZJ1 |
| Gene Alias | 5830461H18Rik; KREV; K-REV; RAP2; RAP2, member of RAS oncogene family (K-rev); Rap2a; Rap2A GTPase; RAP2A, member of RAS oncogene family; RAS related protein 2a; ras-related protein Rap-2a; RbBP-30; RP11-128N14.2 |
| Gene Symbols | RAP2A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.