Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAP2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530144 Supplier Invitrogen™ Supplier No. PA530144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, H1299, HepG2, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (98%), Xenopus laevis (96%), Pig (100%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This intronless gene belongs to a family of RAS-related genes. The proteins encoded by these genes share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. The most striking difference between the RAP and RAS proteins resides in their 61st amino acid: glutamine in RAS is replaced by threonine in RAP proteins. Evidence suggests that this protein may be polyisoprenylated and palmitoylated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP2B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene RAP2B
Gene Accession No. P61225, P61226
Gene Alias 4021402C18Rik; AA408554; Rap2b; RAP2B, member of RAS oncogene family; ras-related protein Rap-2b; small GTP binding protein
Gene Symbols RAP2B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RAP2B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5912, 74012
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.