Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAP30 Antibody (SR1503), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | RAP30 |
---|---|
Clone | SR1503 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
RAP30 Monoclonal antibody specifically detects RAP30 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
RAP30 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ATP-dependent helicase GTF2F2, BTF4, General transcription factor IIF 30 kDa subunit, general transcription factor IIF subunit 2, general transcription factor IIF, polypeptide 2, 30kDa, TF2F2, TFIIF, TFIIF-beta, Transcription initiation factor IIF subunit beta, Transcription initiation factor RAP30 | |
A synthesized peptide derived from RAP30 (Uniprot #: P13984) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1503 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Chromatin Research | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
2963 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title