Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAP74 Antibody (OTI4B10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP317972F
This item is not returnable.
View return policy
Description
RAP74 Monoclonal antibody specifically detects RAP74 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
RAP74 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
OTI4B10 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
BTF4, General transcription factor IIF 74 kDa subunit, general transcription factor IIF subunit 1, general transcription factor IIF, polypeptide 1, 74kDa, TF2F1, TFIIF, TFIIF-alpha, Transcription initiation factor IIF subunit alpha, transcription initiation factor RAP74 | |
Full length human recombinant protein of human GTF2F1(NP_002087) produced in HEK293T cell. | |
0.1 mL | |
Chromatin Research | |
2962 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction