Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAP80 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002346AF700
Description
RAP80 Polyclonal antibody specifically detects RAP80 in Human samples. It is validated for Western BlotSpecifications
RAP80 | |
Polyclonal | |
Western Blot | |
BRCA1-A complex subunit RAP80, RAP80X2HRIP110, receptor associated protein 80, Receptor-associated protein 80, retinoid x receptor interacting protein, Retinoid X receptor-interacting protein 110, RXRIP110, ubiquitin interaction motif containing 1, Ubiquitin interaction motif-containing protein 1 | |
The immunogen recognized by this antibody maps to a region between residue 600 and 650 of human Receptor Associated Protein 80 using the numbering given in entry NP_057374.3 (GeneID 51720). | |
0.1 mL | |
Breast Cancer, Cancer | |
51720 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction