Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAP80 Monoclonal Antibody (A1B11)
GREENER_CHOICE

Catalog No. PIMA536117
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536117 Supplier Invitrogen™ Supplier No. MA536117
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAP80 Monoclonal Antibody for Flow, IHC (P), Western Blot

RAP80 was initially identified as zinc-finger containing nuclear protein that is highly expressed in testis and interacts with the retinoid-related testis-associated receptor (RTR). Later experiments revealed that RAP80 is recruited by the Coiled-coil domain 98 (CCDC98) protein to the breast cancer-1 protein BRCA1, allowing the formation of BRCA1 foci in response to DNA damage caused by ionizing radiation. Both RAP80 and CCDC98 are required for DNA damage resistance, G2-M checkpoint control, and DNA repair. Cells depleted of either RAP80 or CCDC98 exhibited increased sensitivity to ionizing radiation, although not as much as in BRCA1-depleted cells, suggesting that RAP80 and CCDC98 control only part of the DNA damage response role of BRCA1. At least four isoforms of RAP80 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP80
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A1B11
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene UIMC1
Gene Accession No. Q5PQK4, Q5U5Q9, Q96RL1
Gene Alias 9430016E08Rik; BRCA1-A complex subunit RAP80; D330018D10Rik; D630032M02Rik; RAP80; Receptor-associated protein 80; retinoid X receptor interacting protein 110; retinoid X receptor-interacting protein 110; RGD1307009; RIP110; Rxrip110; ubiquitin interaction motif containing 1; ubiquitin interaction motif-containing protein 1; Uimc1; X2HRIP110
Gene Symbols UIMC1
Host Species Mouse
Immunogen Recombinant protein within Human RAP80 aa 330-524.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20184, 290997, 51720
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.