Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAPGEF1 Monoclonal Antibody (OTI2F5)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526280
Description
RAPGEF1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined.
Specifications
RAPGEF1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13905 | |
RAPGEF1 | |
Full length human recombinant protein of RAPGEF1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI2F5 | |
Unconjugated | |
RAPGEF1 | |
4932418O06Rik; C3G; C3G-1; C3G-2; CRK SH3-binding GNRP; GRF2; guanine nucleotide releasing factor 2; Guanine nucleotide-releasing factor 2; guanine nucleotide-releasing factor 2 (specific for crk proto-oncogene); Protein C3G; Rap guanine nucleotide exchange factor (GEF) 1; Rap guanine nucleotide exchange factor 1; Rapgef1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
107746, 2889, 491286, 63881 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction