Learn More
Invitrogen™ RAPGEF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116652
Description
Positive Samples: Mouse liver Immunogen sequence: ARGVAARPGT LHDFHSHEIA EQLTLLDAEL FYKIEIPEVL LWAKEQNEEK SPNLTQFTEH FNNMSYWVRS IIMLQEKAQD RERLLLKFIK IMKHLRKLNN FNSYLAILSA LDSAPIRRLE WQKQTSEGLA EYCTLIDSSS SFRAYRAALS EVEPPCIPYL GLILQDLTFV HLGNPDYIDG KVNFSKRWQQ FNILDSMRCF QQAHYDMRRN DDIINFFNDF SDHLAEEALW ELSLKIKPRN ITRRKTDREE KT.
This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined.
Specifications
| RAPGEF1 | |
| Polyclonal | |
| Unconjugated | |
| RAPGEF1 | |
| 4932418O06Rik; C3G; C3G-1; C3G-2; CRK SH3-binding GNRP; GRF2; guanine nucleotide releasing factor 2; Guanine nucleotide-releasing factor 2; guanine nucleotide-releasing factor 2 (specific for crk proto-oncogene); Protein C3G; Rap guanine nucleotide exchange factor (GEF) 1; Rap guanine nucleotide exchange factor 1; Rapgef1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 107746, 2889, 63881 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.93 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q13905 | |
| RAPGEF1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 844-1095 of human RAPGEF1 (NP_9413721). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.