Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAPGEF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559147
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559147 Supplier Invitrogen™ Supplier No. PA559147
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAVEDSPALG APRVGALPDV VPEGTLLNMV LRRMHRPRSC SYQLLLEHQR PSCIQGLRWT PLTNSEESLD FSESLEQAST ERVLRAGRQL HRHLLATCPN L Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 77%.

Gene ontology (GO) annotation includes small GTPase mediated signal transduction. Target protein binds cAMP and activates the Ras superfamily guanine nucleotide binding protein (Rap1A)in a PKA-independent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAPGEF3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAPGEF3
Gene Accession No. O95398
Gene Alias 2310016P22Rik; 9330170P05Rik; bcm910; CAMP-GEFI; cAMP-regulated guanine nucleotide exchange factor I; cAMP-regulated guanine nucleotide exchange factor I (cAMP-GEFI); CGEF1; Epac; EPAC 1; Epac1; Exchange factor directly activated by cAMP 1; exchange protein directly activated by cAMP 1; HSU79275; LOW QUALITY PROTEIN: rap guanine nucleotide exchange factor 3; MGC21410; Rap guanine nucleotide exchange factor (GEF) 3; Rap guanine nucleotide exchange factor 3; RAP guanine-nucleotide-exchange factor 3; Rap1 guanine-nucleotide-exchange factor directly activated by cAMP; Rapgef3
Gene Symbols RAPGEF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAPGEF3. Recombinant protein control fragment (Product #RP-97655).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10411
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.