Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAPGEF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA129018
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA129018 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA129018 Supplier Invitrogen™ Supplier No. PA129018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-29018 detects Epac1 from human, rat, mouse samples.

Gene ontology (GO) annotation includes small GTPase mediated signal transduction. Target protein binds cAMP and activates the Ras superfamily guanine nucleotide binding protein (Rap1A)in a PKA-independent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAPGEF3
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation Tris glycine with 0.5% BSA, 30% glycerol and 0.02% Sodium azide; pH 7.4-7.9
Gene RAPGEF3
Gene Accession No. O95398, Q8VCC8, Q9Z1C8
Gene Alias 2310016P22Rik; 9330170P05Rik; bcm910; CAMP-GEFI; cAMP-regulated guanine nucleotide exchange factor I; cAMP-regulated guanine nucleotide exchange factor I (cAMP-GEFI); CGEF1; Epac; EPAC 1; Epac1; Exchange factor directly activated by cAMP 1; exchange protein directly activated by cAMP 1; HSU79275; LOW QUALITY PROTEIN: rap guanine nucleotide exchange factor 3; MGC21410; Rap guanine nucleotide exchange factor (GEF) 3; Rap guanine nucleotide exchange factor 3; RAP guanine-nucleotide-exchange factor 3; Rap1 guanine-nucleotide-exchange factor directly activated by cAMP; Rapgef3
Gene Symbols RAPGEF3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 526-541 of Epac1 protein, peptide was covalently modified.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100050541, 10411, 223864, 59326
Target Species Human, Mouse, Rat, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.