Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAPGEF3 Recombinant Rabbit Monoclonal Antibody (JM63-29)
GREENER_CHOICE

Catalog No. PIMA532776
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532776 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532776 Supplier Invitrogen™ Supplier No. MA532776
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Gene ontology (GO) annotation includes small GTPase mediated signal transduction. Target protein binds cAMP and activates the Ras superfamily guanine nucleotide binding protein (Rap1A)in a PKA-independent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAPGEF3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone JM63-29
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene RAPGEF3
Gene Accession No. O95398, Q8VCC8
Gene Alias 2310016P22Rik; 9330170P05Rik; bcm910; CAMP-GEFI; cAMP-regulated guanine nucleotide exchange factor I; cAMP-regulated guanine nucleotide exchange factor I (cAMP-GEFI); CGEF1; Epac; EPAC 1; Epac1; Exchange factor directly activated by cAMP 1; exchange protein directly activated by cAMP 1; HSU79275; LOW QUALITY PROTEIN: rap guanine nucleotide exchange factor 3; MGC21410; Rap guanine nucleotide exchange factor (GEF) 3; Rap guanine nucleotide exchange factor 3; RAP guanine-nucleotide-exchange factor 3; Rap1 guanine-nucleotide-exchange factor directly activated by cAMP; Rapgef3
Gene Symbols RAPGEF3
Host Species Rabbit
Immunogen Synthetic peptide within Human Epac1 aa 874-923.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10411, 223864
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.