Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAPGEF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572746
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572746 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572746 Supplier Invitrogen™ Supplier No. PA572746
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAPGEF4 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Guanine nucleotide exchange factor (GEF) for RAP1A, RAP1B and RAP2A small GTPases that is activated by binding cAMP. Seems not to activate RAB3A. Involved in cAMP-dependent, PKA-independent exocytosis through interaction with RIMS2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAPGEF4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Rapgef4
Gene Accession No. Q8WZA2, Q9EQZ6, Q9Z1C7
Gene Alias 1300003D15Rik; 5730402K07Rik; 6330581N18Rik; cAMP-dependent Rap1 guanine-nucleotide exchange factor; cAMP-GEFII; cAMP-regulated guanine nucleotide exchange factor II; cAMP-regulated guanine nucleotide exchange factor IIB; CAMPS; Cgef2; EPAC; epac 2; Epac2; Epac-2; Exchange factor directly activated by cAMP 2; exchange protein directly activated by cAMP 2; gefii; mKIAA4040; Nbla00496; putative protein product of Nbla00496; Rap guanine nucleotide exchange factor (GEF) 4; Rap guanine nucleotide exchange factor 4; RAP guanine-nucleotide-exchange factor (GEF) 4; Rapgef4
Gene Symbols Rapgef4
Host Species Rabbit
Immunogen EPAC2 antibody for a 15 amino acid synthetic peptide near the amino terminus of human EPAC2. The immunogen is within the first 50 amino acids of EPAC2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11069, 252857, 56508
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.