Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAPGEF4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5115183

Catalog No. PIPA5115183


Only null left
Add to Cart

Description

Description

Antibody detects endogenous levels of total Epac2.

Guanine nucleotide exchange factor (GEF) for RAP1A, RAP1B and RAP2A small GTPases that is activated by binding cAMP. Seems not to activate RAB3A. Involved in cAMP-dependent, PKA-independent exocytosis through interaction with RIMS2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RAPGEF4
Polyclonal
Unconjugated
Rapgef4
1300003D15Rik; 5730402K07Rik; 6330581N18Rik; cAMP-dependent Rap1 guanine-nucleotide exchange factor; cAMP-GEFII; cAMP-regulated guanine nucleotide exchange factor II; cAMP-regulated guanine nucleotide exchange factor IIB; CAMPS; Cgef2; EPAC; epac 2; Epac2; Epac-2; Exchange factor directly activated by cAMP 2; exchange protein directly activated by cAMP 2; gefii; mKIAA4040; Nbla00496; putative protein product of Nbla00496; Rap guanine nucleotide exchange factor (GEF) 4; Rap guanine nucleotide exchange factor 4; RAP guanine-nucleotide-exchange factor (GEF) 4; Rapgef4
Rabbit
Affinity Chromatography
RUO
11069, 252857, 56508
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
Q8WZA2, Q9EQZ6, Q9Z1C7
Rapgef4
A synthesized peptide derived from human RAPGEF4(Accession Q8WZA2), corresponding to amino acid residues N944-D994.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.