Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAPGEF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556019
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556019 Supplier Invitrogen™ Supplier No. PA556019
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VIYGKGVVCT LHEGDDFGKL ALVNDAPRAA SIVLREDNCH FLRVDKEDFN RILRDVEANT VRLKEHDQDV LVLEKVPAGN RASNQGNSQP QQKYTVMSGT PEKILEHFLE TIRLEATLNE ATDSVLNDFI MMHCVFMPNT QLC Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

Guanine nucleotide exchange factor (GEF) for RAP1A, RAP1B and RAP2A small GTPases that is activated by binding cAMP. Seems not to activate RAB3A. Involved in cAMP-dependent, PKA-independent exocytosis through interaction with RIMS2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAPGEF4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rapgef4
Gene Accession No. Q8WZA2
Gene Alias 1300003D15Rik; 5730402K07Rik; 6330581N18Rik; cAMP-dependent Rap1 guanine-nucleotide exchange factor; cAMP-GEFII; cAMP-regulated guanine nucleotide exchange factor II; cAMP-regulated guanine nucleotide exchange factor IIB; CAMPS; Cgef2; EPAC; epac 2; Epac2; Epac-2; Exchange factor directly activated by cAMP 2; exchange protein directly activated by cAMP 2; gefii; mKIAA4040; Nbla00496; putative protein product of Nbla00496; Rap guanine nucleotide exchange factor (GEF) 4; Rap guanine nucleotide exchange factor 4; RAP guanine-nucleotide-exchange factor (GEF) 4; Rapgef4
Gene Symbols Rapgef4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAPGEF4. Recombinant protein control fragment (Product #RP-95156).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11069
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.