Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAPGEF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558193 Supplier Invitrogen™ Supplier No. PA558193
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNCSVDSMSA ALQDERCSSQ ALAVPESTGA LEKTEHASGI GDHSQHGPGW TLLKPSLIKC LAVSSSVSNE EISQEHIIIE AADS Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.

RAPGEF6 is a guanine nucleotide exchange factor (GEF) for Rap1A, Rap2A and M-Ras GTPases. RAPGEF6 does not interact with cAMP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAPGEF6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAPGEF6
Gene Accession No. Q8TEU7
Gene Alias A530068K01; AI844632; BB085664; C030018K18Rik; KIA001LB; LOW QUALITY PROTEIN: rap guanine nucleotide exchange factor 6; mKIAA4052; PDZ domain containing guanine nucleotide exchange factor (GEF) 2; PDZ domain containing guanine nucleotide exchange factor(GEF)1; PDZ domain-containing guanine nucleotide exchange factor 2; PDZ domain-containing guanine nucleotide exchange factor I; PDZGEF2; PDZ-GEF2; RAGEF2; RA-GEF-2; Rap guanine nucleotide exchange factor (GEF) 6; rap guanine nucleotide exchange factor 2; Rap guanine nucleotide exchange factor 6; RAPGEF6
Gene Symbols RAPGEF6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAPGEF6. Recombinant protein control fragment (Product #RP-96914).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51735
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.