Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAPGEF6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558194
Description
Immunogen sequence: FQSLPNPKSW DFLNSYRHTH LDDPIAEVEP TDSEPYSCSK SCSRTCGQCK GSLERKSWTS SSSLSDTYEP NYGTVKRRVL ESTPAESSEG LDPKDATDP Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.
RAPGEF6 is a guanine nucleotide exchange factor (GEF) for Rap1A, Rap2A and M-Ras GTPases. RAPGEF6 does not interact with cAMP.
Specifications
RAPGEF6 | |
Polyclonal | |
Unconjugated | |
RAPGEF6 | |
A530068K01; AI844632; BB085664; C030018K18Rik; KIA001LB; LOW QUALITY PROTEIN: rap guanine nucleotide exchange factor 6; mKIAA4052; PDZ domain containing guanine nucleotide exchange factor (GEF) 2; PDZ domain containing guanine nucleotide exchange factor(GEF)1; PDZ domain-containing guanine nucleotide exchange factor 2; PDZ domain-containing guanine nucleotide exchange factor I; PDZGEF2; PDZ-GEF2; RAGEF2; RA-GEF-2; Rap guanine nucleotide exchange factor (GEF) 6; rap guanine nucleotide exchange factor 2; Rap guanine nucleotide exchange factor 6; RAPGEF6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TEU7 | |
RAPGEF6 | |
Recombinant protein corresponding to Human RAPGEF6. Recombinant protein control fragment (Product #RP-104313). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction