Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAPGEFL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220078
Description
RAPGEFL1 Polyclonal specifically detects RAPGEFL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RAPGEFL1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
Link GEFII, Link guanine nucleotide exchange factor II, Link-GEFII, MGC134798, MGC134799, Rap guanine nucleotide exchange factor (GEF)-like 1, rap guanine nucleotide exchange factor-like 1, RAP guanine-nucleotide-exchange factor (GEF)-like 1 | |
Rabbit | |
73 kDa | |
0.1 mL | |
Neuroscience | |
51195 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q9UHV5 | |
RAPGEFL1 | |
Recombinant protein encompassing a sequence within the center region of human RAPGEFL1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (97%), Bovine (97%), Xenopus tropicalis (82%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction