Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAPGEFL1 Monoclonal Antibody (4G2)
GREENER_CHOICE

Catalog No. PIMA521880
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521880 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521880 Supplier Invitrogen™ Supplier No. MA521880
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MEGPEGLGRK QACLAMLLHF LDTYQGLLQE EEGAGHIIKD LYLLIMKDES LYQGLREDTL RLHQLVETVE LKIPEENQPP SKQVKPLFRH FRRIDSCLQ.

Probable guanine nucleotide exchange factor (GEF).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAPGEFL1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4G2
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RAPGEFL1
Gene Accession No. Q9UHV5
Gene Alias B230105J10; link GEFII; Link guanine nucleotide exchange factor II; Link-GEFII; Rap guanine nucleotide exchange factor (GEF)-like 1; Rap guanine nucleotide exchange factor like 1; rap guanine nucleotide exchange factor-like 1; RAP guanine-nucleotide-exchange factor (GEF)-like 1; Rapgefl1
Gene Symbols RAPGEFL1
Host Species Mouse
Immunogen RAPGEFL1 (NP_057423, 1 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51195
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.