Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAPGEFL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530987
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530987 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530987 Supplier Invitrogen™ Supplier No. PA530987
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa. Predicted reactivity: Mouse (97%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Probable guanine nucleotide exchange factor (GEF).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAPGEFL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene RAPGEFL1
Gene Accession No. Q9UHV5
Gene Alias B230105J10; link GEFII; Link guanine nucleotide exchange factor II; Link-GEFII; Rap guanine nucleotide exchange factor (GEF)-like 1; Rap guanine nucleotide exchange factor like 1; rap guanine nucleotide exchange factor-like 1; RAP guanine-nucleotide-exchange factor (GEF)-like 1; Rapgefl1
Gene Symbols RAPGEFL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RAPGEFL1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51195
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.